Learn More
BRAND™ Spring Clip
Spring Clips designed for wall mounting of cylindrical apparatus components.
£2.30 - £2.90
Specifications
Type Spring Clip
Material Zinc, Polyethylene-Coated Steel
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10097390
View Documents Promotion Details
BRAND™
76045
32 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10488372
View Documents Promotion Details
BRAND™
76055
51 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Spring steel, coated with zinc and polyethylene
  • Non-scratching
  • Good chemical resistance
  • For wall mounting of cylindrical apparatus components like pipes, tubing, etc.
Specifications
Specifications
Spring Clip
Zinc, Polyethylene-Coated Steel