Learn More
GVS SPR491 Elipse Mask with ABEK1P3 Filters
GREENER_CHOICE
Product Code. 18459645
Change view
Click to view available options
Filter Type:
ABEK1P3
Unit Size:
Each
Product Code. Filter Type unitSize
18459645 ABEK1P3 Each
1 options
Product Code. 18459645 Supplier GVS Supplier No. SPR491
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This mask is recommended for multigas and dust risks, in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
This mask is recommended for multigas and dust risks, in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
  • Extremely lightweight and compact to give the wearer as much comfort as possible. It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
  • Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
  • Filters last up to 3 years and can be easily replaced.
  • Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
  • Certifications: Mask Body EN140: 1998. ABEK1P 3 R D Filters EN14387: 2004 + A1: 2008. CE 2797
Specifications
Color Gray
Disposable Reusable
Filter Type ABEK1P3