missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Spouted Mortar

Brand:  Haldenwanger™ 55/6a/rauh

Code : 02

Additional Details : Weight : 1.16000kg

 View more versions of this product

Product Code. 12045143

  • £53.95 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

700mL
Mortar
Mortar
150mm
With spout
Videos