missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Spouted Mortar

Brand:  Haldenwanger™ 55/1/glasiert

Code : 02

Additional Details : Weight : 0.27000kg

 View more versions of this product

Product Code. 12035133

  • £25.01 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

75mL
Mortar
Mortar
70mm
With spout
Videos