missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Spouted Mortar

Brand:  Haldenwanger™ 55/2/glasiert

Code : 02

Additional Details : Weight : 0.31000kg

 View more versions of this product

Product Code. 12055133

  • £41.35 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

100mL
Mortar
Mortar
80mm
With spout