missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Spouted Mortar

Brand:  Haldenwanger™ 55/4/rauh

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12005143

  • £37.17 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Mortar
300mL
With spout
Mortar
115mm