missing translation for 'onlineSavingsMsg'
Learn More

Sponge Nickel, A-5000, nonpromoted, Alfa Aesar™

Slurry in water

Brand:  Alfa Aesar 044867.22

Code : NEW

Additional Details : CAS Number : 7440-02-0 Weight : 0.10000kg Transport : UN number : 1378 Chem class : 1 Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 11386279

  • £33.40 / 100g


Estimated Shipment
23-01-2018
Add to basket

Specifications

Chemical Identifiers

Nickel
Sponge A-5000
MFCD00167392
Nonpromoted
Ni

Specifications

Nickel
Nonpromoted
8.908g/cm3 at 20°C
Ni
Odorless
100g
Silver colored sponge
Sponge A-5000
MFCD00167392
Silver
Videos
SDS