Learn More
SPOCK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552435
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FKDGKLSNNE WCYCFQKPGG LPCQNEMNRI QKLSKGKSLL GAFIPRCNEE GYYKATQCHG STGQCWCVDK YGNELAGSRK QGAVSCEEEQ ETSGDFGSGG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
SPOCK1 Polyclonal antibody specifically detects SPOCK1 in Human samples. It is validated for Immunohistochemistry.Specifications
SPOCK1 | |
Polyclonal | |
Unconjugated | |
SPOCK1 | |
FLJ37170; Protein SPOCK; sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1; sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1; Spock; Spock1; testican; testican 1; testican-1; TIC1; TICN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08629 | |
SPOCK1 | |
Recombinant protein corresponding to Human SPOCK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only