Learn More
SPOCD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556828
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GLNIIEQQQK EPCRLPASKM THKGEVEIQR DMDQTLTLED LVGPQMFMDC SPQALPIASE DTTGQHDHHF LDPNCHICKD WEPSNELLGS FEAAKSC Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 31%.
SPOCD1 Polyclonal antibody specifically detects SPOCD1 in Human samples. It is validated for Immunohistochemistry.Specifications
SPOCD1 | |
Polyclonal | |
Unconjugated | |
SPOCD1 | |
PPP1R146; protein phosphatase 1, regulatory subunit 146; SPOC domain containing 1; SPOC domain-containing protein 1; SPOCD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMY3 | |
SPOCD1 | |
Recombinant protein corresponding to Human SPOCD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only