missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Thermo Scientific Splitless Liners with Single Taper

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09214401

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12964051

  • £36.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Thermo Scientific instruments
Specifications

Specifications

1Each
5mm
105mm
GC Inlet Liner
8mm
Videos