Learn More
Nabertherm™ Split Type Horizontal Tube Furnace RSH 1300 Series with P580 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
£6496.68 - £12666.38
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17338159
|
Nabertherm™
RS252W3AN3 |
50 mm | 500 mm |
£6496.68
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17378159
|
Nabertherm™
RS322W3AN3 |
120 mm | 500 mm |
£8105.79
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17318169
|
Nabertherm™
RS382W3AN3 |
80 mm | 500 mm |
£7873.46
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17358169
|
Nabertherm™
RS422W3AN3 |
120 mm | 750 mm |
£10308.64
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17398169
|
Nabertherm™
RS472W3AN3 |
170 mm | 750 mm |
£11332.62
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17338179
|
Nabertherm™
RS482W3AN3 |
80 mm | 750 mm |
£9938.63
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17378179
|
Nabertherm™
RS522W3AN3 |
120 mm | 1000 mm |
£11237.97
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17318189
|
Nabertherm™
RS572W3AN3 |
170 mm | 1000 mm |
£12666.38
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1300°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type S thermocouple
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 including fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- Switchgear and control unit integrated in furnace housing
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
P580 | |
Up to 1300°C |