missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ PerkinElmer Split / Splitless FocusLiners
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09209225
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with PerkinElmer instruments
Specifications
Split/Splitless FocusLiner | |
92 mm | |
6.2 mm |
25/Pk. | |
4 mm |