Learn More
Spinel Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific™
100 Orientation
Brand: Thermo Scientific Alfa Aesar 038505.KU
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Spinel Substrate | |
Plate | |
MgAl2O4 | |
Magnesium aluminum oxide |
Polished One Side | |
5 Ea. | |
MFCD00049475 |