missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPIN4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550265
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SPIN4 protein.
SPIN4 Polyclonal antibody specifically detects SPIN4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SPIN4 | |
Polyclonal | |
Unconjugated | |
SPIN4 | |
9630042H07Rik; FLJ44984; MGC133224; novel Spin/Ssty repeat containing protein; Spin4; spindlin family member 4; spindlin family, member 4; spindlin-4; TDRD28 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
139886 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q56A73 | |
SPIN4 | |
Full length fusion protein of Human SPIN4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |