Learn More
New Pig™ Spill kit refill for kit 328 for acids, caustics and unknown solvents 56 x 43 x 410mm absorbs up to 42L pink
Brand:  New Pig™ RFL328
Code : 04
Additional Details : Weight : 1.00000kg
Product Code. 12953585