missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPI-B Mouse anti-Human, Biotin, Clone: 235D, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-9790-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
The 235D monoclonal antibody recognizes human SPI-B. SPI-B is a member of the ETS transcription factor family which is comprised of DNA binding proteins that function in lymphoid development and activity. SPI-B binds the PU box, a purine-rich DNA sequence (5’-GAGGAA-3’). SPI-B is expressed in mature B cells, immature T cells, plasmacytoid dendritic cells, and microfold cells (M cells) of the intestine. In human tonsil tissue, high levels of SPI-B are expressed in plasmacytoid dendritic cells, while lower levels of expression are found in germinal center B cells.Specifications
SPI-B | |
235D | |
Biotin | |
Conjugated | |
Murine | |
100μg | |
Store at 2-8°C. Do not freeze. | |
Monoclonal |
Microscopy | |
0.5mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
ETS | |
IgG1, kappa | |
RUO | |
Primary | |
Human |
For Research Use Only.