Learn More
Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets
Keep spare items to ensure there is no down time.
Brand: Cole-Parmer™ 6881C20
Code : NEW
Additional Details : Weight : 5.00000kg
Requires : 886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
Description
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
Center Cylinder | |
Polycarbonate | |
20 Pk. |
6881 Vial Set | |
Cole-Parmer Freezer/Mill |