Learn More
SPEX™ SamplePrep Geno/Grinder™ Accessory, Grinding Mill
Compatible with all SPEX SamplePrep Titer Plates and Vial Sets
Brand: SPEX™ SamplePrep 2166
Code : 50
Additional Details : Weight : 0.50000kg
Description
- Acid washed silica grinding beads
- Bead size ranges from 800-1000μm
- Suitable for basic cell lysis applications using the 2010 Geno/Grinder
- Treated to remove fine particles and contaminates
Specifications
Silica Grinding Beads | |
Titer plates and vial sets of Geno/Grinder | |
400 to 600 μm |
Ideal for basic cell lysis | |
Silica |