Learn More
Cole-Parmer™ Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer
These Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
Brand: Cole-Parmer™ 2661
Code : 46
Additional Details : Weight : 5.00000kg
Description
- Safely grind samples in the HG-600 Geno/Grinder™ 2010.
- Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
- Order homogenizer and vials separately.
Specifications
Holder | |
Spex Geno/Grinder | |
Aluminum |
Ideal for High Throughput Cryogenic Grinding and to Preserve Pesticides, RNA, Proteins and Other Temperature Sensitive Samples | |
15 x 15 mL Polycarbonate Cryovials, each pair holds up to 30 cryovials |