Learn More
SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards
SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the
QuEChERS analytical techniques (GC, GC/MS, LC/MS)
Brand: SPEX CertiPrep™ QUECR-PCB-28
Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
Single Component Reference Standards
- Volume: 5mL
Specifications
50 μg/mL | |
PCB 28 | |
5 mL |
QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) | |
QuEChERS standard- PCB 28 | |
Acetonitrile |