Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
Brand:  SPEX CertiPrep™ AOAC-PSA-15ML
Code : 04
Additional Details : Weight : 1.18000kg Transport : UN number : 3264 Chem class : 1a Pack group : III
 View more versions of this product
Product Code. 11475908
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Sensible solutions for pesticide extraction
AOAC 2007.01
50 pk.
15mL
400mg PSA, 1200mg MgSO4
PSA dispersive SPE clean-up tube
400mg PSA, 1200mg MgSO4
Videos