Learn More
SPEX CertiPrep™ Method AOAC 2007.01 SPEXQuE™ QuEChERS Kit
Sensible Solutions for Pesticide Extraction
Brand:  SPEX CertiPrep™ AOAC-ACE-50ML
Code : 04
Additional Details : Weight : 0.27000kg Transport : UN number : 3264 Chem class : 1a Pack group : III
 View more versions of this product
Product Code. 11465908
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
  • US (AOAC Method 2007.01)
Specifications
Specifications
Sensible solutions for pesticide extraction
AOAC 2007.01
50 pk.
50mL
6g MgSO4, 1.5g sodium acetate
Acetate buffered extraction tube
6g MgSO4, 1.5g Sodium Acetate
Videos