missing translation for 'onlineSavingsMsg'
Learn More

Spermine NONOate, Tocris Bioscience™

Slow NO releasing agent

Brand:  Tocris (Bio-Techne) 113510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488703

  • £92.00 / 10mg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

A stable complex of nitric oxide and spermine used to generate a controlled release of nitric oxide in solution (EC50 = 6.2 μM for relaxation of rabbit aorta, and t½ =39 min at 37°C, pH 7.4, aqueous solution).
Specifications

Chemical Identifiers

Spermine NONOate
C10H26N6O2
262.35
N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine

Specifications

Spermine NONOate
>98%
262.35
10mg
136587-13-8
C10H26N6O2
N-[4-[1-(3-Aminopropyl)-2-hydroxy-2-nitrosohydrazino]butyl-1, 3-propanediamine
Videos