missing translation for 'onlineSavingsMsg'
Learn More

Spermine, 97%, Thermo Scientific Chemicals

Catalog Number 11385325
Click to view available options
Quantity:
1 g
5 g
71-44-3
C10H26N4
202.35
MFCD00008215
PFNFFQXMRSDOHW-UHFFFAOYSA-N
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
1103
CHEBI:15746
N,N'-bis(3-aminopropyl)butane-1,4-diamine
NCCCNCCCCNCCCN
71-44-3
C10H26N4
202.35
MFCD00008215
PFNFFQXMRSDOHW-UHFFFAOYSA-N
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
1103
CHEBI:15746
N,N'-bis(3-aminopropyl)butane-1,4-diamine
NCCCNCCCCNCCCN

Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

Solubility
Soluble in chloroform, methanol, water and lower alcohols. Insoluble in ether, benzene and petroleum ether.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 28°C to 30°C
Boiling Point 150°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3259
Beilstein 1751791
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,8743
Solubility Information Soluble in chloroform,methanol,water and lower alcohols. Insoluble in ether,benzene and petroleum ether.
Formula Weight 202.35
Percent Purity 97%
Chemical Name or Material Spermine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'