missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermidine Trihydrochloride, Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Description
Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms
Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins
Spermidine Trihydrochloride forms stable compounds with nucleic acids. It is used in biochemical research.
Specifications
Melting Point | 248°C |
Color | White |
pH | 3 to 6 |
Assay Percent Range | ≥99 % |
Packaging | Amber Glass |
Quantity | 1 g |
Merck Index | 15, 8869 |
Formula Weight | 254.63 |
Percent Purity | 99% min. |
Physical Form | Solid |
Show More |
Safety and Handling
Product Identifier
- Spermidine trihydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'