missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, 99%, Thermo Scientific Chemicals

Catalog Number 10607934
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
124-20-9
C7H19N3
145.25
MFCD00008229
ATHGHQPFGPMSJY-UHFFFAOYSA-N
spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
1102
CHEBI:16610
NCCCCNCCCN
124-20-9
C7H19N3
145.25
MFCD00008229
ATHGHQPFGPMSJY-UHFFFAOYSA-N
spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
1102
CHEBI:16610
NCCCCNCCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 22.0°C to 25.0°C
Color Colorless
Density 0.9250g/mL
Boiling Point 128.0°C to 130.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4785 to 1.4805
Quantity 25 g
Beilstein 04,II,704
Merck Index 15,8869
Specific Gravity 0.925
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
IUPAC Name (4-aminobutyl)(3-aminopropyl)amine
Formula Weight 145.25
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material Spermidine
Show More Show Less
compliance-icons
Product Identifier
  • Spermidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'