missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ MediaKap™ and MediaKap Plus HF Filters
0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling
Brand: Spectrum™ MP2M 10B 12S
Code : LB
Additional Details : Weight : 0.01000kg
Description
- Efficiently sterilize and clarify culture medium or buffer solution
- Smaller, narrow design for low hold-up volume
- Polypropylene vent prevents air lock
- Operates with peristaltic pump, vacuum, or syringe
- Available in five different sizes that filter designated volume in 15 to 20 minutes
- Sterile, biocompatible and nonpyrogenic
- USP Class VI; 100% integrity tested
- Optional filling bell maintains sterility while filling bottles
- MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium
Specifications
0.25HB/0.25HB | |
Polycarbonate, Polyurethane | |
0.2 μm | |
100 cm2 | |
Efficiently sterilize and clarify culture medium or buffer solution | |
12 Case | |
5 to 10 L |
MediaKap-10 Plus | |
MCE | |
Sterile | |
1000 mL/min. at 10psig | |
Automatic vent to prevent air lock, filling bell | |
Bottle Top Filter |