missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ MediaKap™ and MediaKap Plus HF Filters
0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling
£147.00 - £780.00
Specifications
Material | Polycarbonate, Polyurethane |
---|---|
Material (Membrane) | MCE |
Pore Size | 0.2 μm |
Sterility | Sterile |
For Use With (Application) | Efficiently sterilize and clarify culture medium or buffer solution |
Product Code | Brand | Connectors | Filter Type | Includes | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connectors | Filter Type | Includes | Volume (Metric) | Price | Quantity & Availability | ||
10176794
|
Spectrum™
ME2M-020-18S |
Female luer lok/0.25HB | MediaKap-2 | Automatic vent to prevent air lock | 0.2 to 2 L |
£438.00
Pack of 18 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
11745495
|
Spectrum™
ME2M-05B-12S |
Female luer lok/male luer slip | MediaKap-5 | Automatic vent to prevent air lock, filling bell | 2 to 5 L |
£455.00
Pack of 12 |
Estimated Shipment: 09-05-2024
Log in to see stock. |
||
11486988
|
Spectrum™
ME2M-10B-12S |
0.25HB/0.25HB | MediaKap-10 | Automatic vent to prevent air lock, filling bell | 5 to 10 L |
£780.00
Pack of 12 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
11416998
|
Spectrum™
ME2M-25B-06S |
VarHB/VarHB | MediaKap-25 | Automatic vent to prevent air lock, filling bell | 10 to 25 L |
£535.00
Pack of 6 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
11426998
|
Spectrum™
ME2M-50B-03S |
VarHB/VarHB | MediaKap-50 | Automatic vent to prevent air lock, filling bell | 25 to 50 L |
£386.00
Pack of 3 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
12811403
|
Spectrum™
ME2M-020-18S |
Female luer lok/0.25HB | MediaKap-2 | Automatic vent to prevent air lock | 0.2 to 2 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11496988
|
Spectrum™
ME2M 100 18S |
0.25HB/0.25HB | MediaKap-10 | Automatic vent to prevent air lock | 5 to 10 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11446998
|
Spectrum™
MP2M 50B 03S |
VarHB/VarHB | MediaKap-50 Plus | Automatic vent to prevent air lock, filling bell | 25 to 50 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11717723
|
Spectrum™
MP2M02018S |
Female luer lok/0.25HB | MediaKap-2 Plus | Automatic vent to prevent air lock | 0.2 to 2 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11737754
|
Spectrum™
MP2M-050-18S |
Female luer lok/0.25HB | MediaKap-5 Plus | Automatic vent to prevent air lock | 2 to 5 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11436998
|
Spectrum™
MP2M 25B 06S |
VarHB/VarHB | MediaKap-25 Plus | Automatic vent to prevent air lock, filling bell | 10 to 25 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11406998
|
Spectrum™
MP2M 10B 12S |
0.25HB/0.25HB | MediaKap-10 Plus | Automatic vent to prevent air lock, filling bell | 5 to 10 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12627943
|
Spectrum™
ME2M-02B-12S |
Female luer lok/0.25HB | MediaKap-2 | Automatic vent to prevent air lock, filling bell | 0.2 to 2 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Efficiently sterilize and clarify culture medium or buffer solution
- Smaller, narrow design for low hold-up volume
- Polypropylene vent prevents air lock
- Operates with peristaltic pump, vacuum, or syringe
- Available in five different sizes that filter designated volume in 15 to 20 minutes
- Sterile, biocompatible and nonpyrogenic
- USP Class VI; 100% integrity tested
- Optional filling bell maintains sterility while filling bottles
- MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium
Specifications
Polycarbonate, Polyurethane | |
0.2 μm | |
Efficiently sterilize and clarify culture medium or buffer solution |
MCE | |
Sterile | |
Bottle Top Filter |