missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ MediaKap™ and MediaKap Plus HF Filters

0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling

Brand:  Spectrum™ ME2M-02B-12S

Code : LB

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12627943

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Efficiently sterilize and clarify culture medium or buffer solution
  • Smaller, narrow design for low hold-up volume
  • Polypropylene vent prevents air lock
  • Operates with peristaltic pump, vacuum, or syringe
  • Available in five different sizes that filter designated volume in 15 to 20 minutes
  • Sterile, biocompatible and nonpyrogenic
  • USP Class VI; 100% integrity tested

  • Optional filling bell maintains sterility while filling bottles
  • MediaKap Plus filters utilize an advanced DynaFibre membrane that further increases filtration efficiency and dramatically reduces the time required to filter serum-enriched medium

Specifications

Specifications

Female luer lok/0.25HB
Polycarbonate, Polyurethane
0.2 μm
35 cm2
Efficiently sterilize and clarify culture medium or buffer solution
12 Case
0.2 to 2 L
MediaKap-2
MCE
Sterile
400 mL/min. at 10psi
Automatic vent to prevent air lock, filling bell
Bottle Top Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers