missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 300 kd

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

Brand:  Spectrum™ 131456T

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15340682

  • £277.00 / 1m
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

Description

Description

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing (16mm flat width), a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water. Nine different MWCOs avilalable: 0.1-0.5 kD, 0.5-1.0 kD, 3.5-5 kD, 8-10 kD, 20 kD, 50 kD, 100 kD, 300 kD and 1000 kD.

  • Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
  • 9 MWCOs available ranging from 100 to 1,000,000 Daltons
  • Ultrapure, no heavy metal or sulfiide impurities
  • Low protein binding
  • No glycerin removal, cleaning or pre-treatment required

Specifications

Specifications

300 kDa
20 mm
3.1 mL/cm vol./length
1 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers