missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Trial Size Kits for Biotech-Grade RC Dialysis Membrane Tubing
0.5 meter (1.6 ft) dialysis tubing length ideal for small applications or membrane evaluation
£109.69
Specifications
Product Type | Dialysis Trial Kit |
---|---|
Diameter (Metric) | 10 mm |
Diameter (English) | 0.39 in. |
Kit Contents | 0.5m Biotech RC tubing with glycerine, 2 universal tubing closures, 5 opening picks |
Quantity | Each |
Includes: 0.5m Biotech RC Tubing with glycerine, 2 Universal Tubing Closures, 5 opening picks
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
15330792
|
Spectrum™
133 342T |
20 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15360782
|
Spectrum™
133 198T |
3.5 to 5 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15300792
|
Spectrum™
133 270T |
8 to 10 kDa |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Ultrapure Biotech RC is a premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
- Properties: Hydrophilic, symmetric porosity
- 3 MWCOs: 3.5-5 kD, 8-10 kD and 20 kD
- Ultrapure, contains no heavy metals or sulfides
Specifications
Dialysis Trial Kit | |
0.39 in. | |
Each |
10 mm | |
0.5m Biotech RC tubing with glycerine, 2 universal tubing closures, 5 opening picks | |
Ideal for membrane and application evaluation for more critical separations |