missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs, 12-14kD MWCO
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs are for basic dialysis and are hydrophilic with symmetric porosity.
£195.12 - £198.28
Specifications
Product Type | Dialysis Membrane Discs |
---|---|
MWCO | 12 to 14 kDa |
Quantity | 50/Pk. |
For Use With (Application) | Basic dialysis |
Code produit | Marque | Diameter (Metric) | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Diameter (Metric) | Prix | Quantité et disponibilité | |||||
11571010
|
Spectrum Labs™
132 498 |
33 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11561010
|
Spectrum Labs™
132 496 |
47 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Dry Standard RC dialysis membrane in single-layer discs.
- Used in various commercially available dialysis equipment.
- Discs are separated by gylcine paper
- Packaged dry with glycerin (rinse out with water)
Spécification
Dialysis Membrane Discs | |
12 to 14 kDa | |
50/Pk. | |
Basic dialysis |