missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Sulfate, ≥98%, MP Biomedicals™

Catalog Number p-4405411
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
23312-56-3
C14H26N2O11S
430.43
MFCD00270185
XGBFWQUQYQIFLB-MTTMTQIXSA-N
U 18409E, antibiotic
CHEBI:9216
(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
This item is not returnable. View return policy
23312-56-3
C14H26N2O11S
430.43
MFCD00270185
XGBFWQUQYQIFLB-MTTMTQIXSA-N
U 18409E, antibiotic
CHEBI:9216
(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
Type Spectinomycin Sulfate
Physical Form Powder
Assay Percent Range ≥98%
PubChem CID 64771
Formula Weight 430.4
Color White
Quantity 5 g
Chemical Name or Material Spectinomycin Sulfate
compliance-icons compliance-icons
Product Identifier
  • Spectinomycin Sulfate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'