missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Specific Gravity Hydrometer, 305 mm

The Precision Specific Gravity Hydrometer is calibrated for use at 60°F (15.55°C).

Brand:  VEE GEE 6603-1

 View more versions of this product

Product Code. 17866513

  • £29.65 / Each
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The specific gravity range and subdivisions differ depending on the model.
Specifications

Specifications

Specific Gravity Hydrometer
30.5 cm
Glass
1.000 to 1.220
0.002
35 x 3 x 3 cm
6 g
60°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers