missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Specific Gravity Hydrometer, 305 mm
The Precision Specific Gravity Hydrometer is calibrated for use at 60°F (15.55°C).
Brand: VEE GEE 6603-7
This item is not returnable.
View return policy
Description
- The specific gravity range and subdivisions differ depending on the model.
Specifications
Specific Gravity Hydrometer | |
30.5 cm | |
Glass | |
1.000 to 2.000 |
0.01 | |
35 x 3 x 3 cm | |
6 g | |
60°F |