missing translation for 'onlineSavingsMsg'
Learn More

SPE tube alumina normal phase polypropylene 1g 6mL Bakerbond

Brand:  JT Baker 7214-07

Code : Z3

Additional Details : CAS Number : 1344-28-1 Weight : 0.10000kg

Product Code. 13154644

  • POA / Pack of 30
Estimated Shipment: 05-06-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.