Learn More
SPDYA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564267
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CCEEVMAIAP THYIWQRERS VHHSGAVRNY NRDEVQLPRG PSATPVDCSL CGKKRRYVRL GLSSSSS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.
SPDYA Polyclonal antibody specifically detects SPDYA in Human samples. It is validated for Immunohistochemistry.Specifications
SPDYA | |
Polyclonal | |
Unconjugated | |
SPDYA | |
4921517J08Rik; 4930548B21Rik; Gs4; hSpy/Ringo A; Lm23; MLZ-465; mSpy/Ringo A; protein expressed in male leptotene and zygotene spermatocytes 465; Rapid inducer of G2/M progression in oocytes A; RINGO A; RINGO3; RINGOA; SPDY1; SPDYA; spdya {ECO:0000250; speedy A; speedy A1; speedy A2; speedy homolog 1; speedy homolog A; speedy homolog A (Xenopus laevis); speedy protein A; speedy/ringo; speedy/RINGO cell cycle regulator family member A; Speedy-1; spermatogenesis related protein Gs4; spy1; UniProtKB:Q5MJ70} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245711 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5MJ70 | |
SPDYA | |
Recombinant protein corresponding to Human SPDYA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only