missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abritech Spatula combustion (heating spoon) nickel-plated rod with 14mm dia bowl 158mm length
Brand: Abritech SMJ-950-T
Spatula combustion (heating spoon) nickel-plated rod with 14mm dia bowl 158mm length
Code : 02
Additional Details : Weight : 0.04324kg