missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA5L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550831
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SPATA5L1 protein.
SPATA5L1 Polyclonal antibody specifically detects SPATA5L1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SPATA5L1 | |
Polyclonal | |
Unconjugated | |
SPATA5L1 | |
AV141009; C130039A10Rik; SPATA5L1; spermatogenesis associated 5 like 1; spermatogenesis associated 5-like 1; spermatogenesis-associated protein 5-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79029 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BVQ7 | |
SPATA5L1 | |
Synthetic peptide corresponding to residues near the C terminal of human spermatogenesis associated 5-like | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |