missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550276
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SP / TFF2 protein.
TFF2 Polyclonal antibody specifically detects TFF2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TFF2 | |
Polyclonal | |
Unconjugated | |
Tff2 | |
mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7032 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q03403 | |
Tff2 | |
Fusion protein corresponding to a region derived from internal residues of human trefoil factor 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |