missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Model 1.0
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets block 99% of UV light and are resistant to staining, crazing and chemical attack.
Brand: Bel-Art™ F42071-0006
Code : 03
Additional Details : Weight : 1.00000kg
Description
- Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Stack up to 3 units high to save space
- Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications
Not Autoclavable | |
Vertical | |
Left-hinged and gasketed | |
Carrying handles, one latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
1 | |
Fixed Perforated | |
16.3 x 13.4 in. (41.4 x 34.1 cm) | |
41.4 cm | |
8.9 in. | |
22.5 cm | |
4.5 in. | |
16.3 in. |
Clear with Blue End Caps | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 | |
1.0 | |
Chemicals, stains and fine cracks | |
0.75 cu. ft. | |
16.3 in. | |
13.4 in. | |
34.1 cm | |
8.4 in. | |
21.3 cm | |
41.4 cm |