missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Bel-Art™ SP Bel-Art Safety-Labeled 4-Color Wide-Mouth Wash Bottles
Click to view available options
Chemical Label:
Distilled Water
Methanol
Sodium Hypochlorite (Bleach)
Capacity (Metric):
500 mL
1000 mL
Description
- Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm; Toluene is red LDPE bottle
- Color-coded polypropylene closures aid in quick and easy identification
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- 500 mL - 4/bag, 1000 mL - 4/bag; assortment is 6 per bag, includes one each: Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB
Specifications
Closure Material | Polypropylene |
Chemical Label | Distilled Water |
Capacity (Metric) | 1000 mL |
Material | Polyethylene |
No. per Pack | 4/Pk. |
Closure Color | Blue |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'