Sox9 Mouse anti-Human, Mouse, Alexa Fluor 488, Clone: GMPR9, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 53-9765-82

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15560287

Quantity Price
1 £ 176.7 / 100µg
Estimated Shipment date
from Supplier 04-05-2017
Add to basket


Description and Specification

Specification

Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Host Species Mouse
Isotype IgG1
Quantity 100μg
Format Conjugated
Clone GMPR9
Species Reactivity Human, Mouse
Primary or Secondary Primary
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Antigen Sox9
Concentration 0.5mg/mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Alexa Fluor 488

This GMPR9 monoclonal antibody reacts with human and mouse Sox9, a member of the Sry HMG-box (Sox) gene family of transcription factors. Like Sry, Sox9 contains a high mobility group (HMG-box) DNA-binding domain. The Sox9 DNA-binding domain recognizes the sequence AGAACAATGG, and when bound to co-factors, functions to bend the DNA. This structural change is thought to be important for chromatin remodeling and the initiation of more broad transcriptional regulation of multiple genes. Sox9 is expressed in multiple tissue types during development and its activation of downstream targets is determined by interaction with tissue-specific co-factors. During development, Sox9 expression is important for cartilage formation (mesenchymal to chondrocyte), sex determination (testes formation), neural crest formation and retinal development. Sox9 plays a role in the differentiation of multiple cell types including glial, hair, cardiac, melanocytes, pancreatic, prostate, and intestinal epithelium. Sox9 regulates expression of the following family member genes sox5, sox6, and sox10, in addition to collagen type 2 alpha, collagen 9A1, CEACAM1, and snail2. Sox9 dysregulation is involved in craniofacial defects, cleft palate, sex reversal, and some types of cancer (melanoma, lung adenocarcinoma, and breast cancer). Sox9 has also been shown to be expressed in adult stem cells in the liver, pancreas and intestine.