missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566194
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PRRRKSAKAGH SDSDSGAELG PHPGGGAVYK AEAGLGDGHH HGDHTGQTH Highest antigen sequence identity to the following orthologs - mouse 73%, rat 70%.
SOX8 Polyclonal antibody specifically detects SOX8 in Human samples. It is validated for Immunocytochemistry.Specifications
SOX8 | |
Polyclonal | |
Unconjugated | |
SOX8 | |
SOX8; Sox-8; SRY (sex determining region Y)-box 8; SRY box 8; SRY-box 8; SRY-box containing gene 8; transcription factor SOX-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57073 | |
SOX8 | |
Recombinant Human SOX8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only