missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565893
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: TLAHLLPAPAM YSLLETELKN PVGTPTQA Highest antigen sequence identity to the following orthologs - mouse 93%, rat 41%.
This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.Specifications
SOX3 | |
Polyclonal | |
Unconjugated | |
SOX3 | |
GHDX; LOC679158; MRGH; PHP; PHPX; similar to SRY (sex determining region Y)-box 3; Sox3; Sox-3; SOXB; SRY (sex determining region Y)-box 3; SRY box 3; SRY-box 3; SRY-box containing gene 3; transcription factor SOX-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P41225 | |
SOX3 | |
Recombinant Human SOX3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only