Learn More
SOX11 Monoclonal Antibody (SOX11-C1), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£85.25 - £240.00
Specifications
Clone | SOX11-C1 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Biotin |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15557386
|
Affymetrix eBioscience
13-9773-82 |
100 μg |
£240.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15547386
|
Affymetrix eBioscience
13-9773-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody SOX11-C1 (C1) recognizes human Sox11. The transcription factor Sox11 is a member of the SOX (sex determining region Y-related HMG (High Mobility Group) Box) family of proteins. Under normal conditions Sox11 is expressed in the developing central nervous system during embryogenesis and has sequence homology to Sox4. Sox11 plays a role in neuronal maturation and epithelial-mesenchymal interactions and is required for the survival of neuronal and mesenchymal progenitor cells. Sox11 is expressed in mantle cell lymphoma (MCL) and in subsets of hairy cell leukemias, Burkitt lymphomas, and B cell lymphoblastic leukemias, but is not expressed in other B cell lymphomas or in normal B lymphocytes. Sox11 is also expressed in epithelial ovarian cancer and gliomas. Applications Reported: This SOX11-C1 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This SOX11-C1 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded tissue using high pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.Specifications
SOX11-C1 | |
Monoclonal | |
Liquid | |
RUO | |
P35716 | |
SOX11 | |
Primary | |
Antibody |
Immunohistochemistry (Paraffin) | |
Biotin | |
Mouse | |
Human | |
6664 | |
IgG1 κ | |
Affinity chromatography | |
SOX11 |
For Research Use Only.