missing translation for 'onlineSavingsMsg'
Learn More

SOX10 Mouse anti-Human, Mouse, Rat, Clone: 20B7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5923-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15587456

  • £301.00 / 100µg

Estimated Shipment
08-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Specifications

Specifications

SOX10
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O55170, Q04888, P56693
SRY, sex determining region Y-box 10 , DOM, MGC15649, WS4, SRY-related HMG-box gene 10
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
20B7
Unconjugated
SOX10
Liquid
Sox10
IgG1, kappa
100 μg
4° C
Primary
20665, 29361, 6663
Videos
Documents

For Research Use Only.