missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Sound Plate

Brand:  Haldenwanger™ Clay Plate 180 mm

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.24800kg

 View more versions of this product

Product Code. 12011842

  • £10.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

180mm