missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Sound Plate

Brand:  Haldenwanger™ Clay plate 220 mm

Code : 02

Additional Details : Weight : 0.39500kg

 View more versions of this product

Product Code. 12021842

  • £12.06 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

220mm
Videos