Learn More
Tocris Bioscience™ Sotalol hydrochloride
Brand:  Tocris Bioscience™ 095250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13488193
  • £220.00 / 50mg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
A relatively potent pure β adrenergic antagonist, unique in possessing additional class III antiarrhythmic activity. Also available as part of the Mixed Adrenergic Tocriset™ .
Specifications
Specifications
N-[4-[1-Hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide hydrochloride
>99%
Observed MW: 308.82
Soluble to 50mM in water
959-24-0
C12H20N2O3S.HCl